Learn More
Candida albicans, FITC, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA127381
Catalog No. PA127381
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Candida albicans is the most frequently isolated fungal pathogen of humans, affecting immuocompromised patients ranging from premature infants to AIDS sufferers. Systemic infections have an attributed mortality of 30-50%. C. albicans is a diploid organism which has eight sets of homologous chromosomes. Its genome size is about 16 Mb (haploid), about 30% greater than S. cerevisiae (baker's yeast).
PA1-27381 detects numerous proteins from a soluble C. albicans extract (IEP) from yeast samples. PA1-27381 has been successfully used in double immunodiffusion, ELISA, and immunocytochemistry procedures.The PA1-27381 immunogen is Candida albicans, type A (ATCC #32354).
Specifications
Specifications
Candida albicans
Polyclonal
FITC
Purified
Rabbit
IgG
100μL
Store at -20°C
Primary
Yeast
ELISA, Immunocytochemistry, Immunodiffusion, Immunoelectrophoresis, Immunofluorescence
4mg/mL
PBS with 10mg/mL BSA and 0.1% sodium azide; pH 7.2
C. albicans
Candida albicans, type A.
Protein A
RUO
Antibody
Polyclonal