Learn More
Candida albicans Rabbit anti-Fungi, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA17206
Catalog No. PA17206
Add to cart
Description
Description
PA1-7206 detects Candida albicans. PA1-7206 has been successfully used in ELISA, immunohistochemistry, immunoblot, and immunofluorescence procedures. The PA1-7206 immunogen is Candida albicans, type A. Recommended short term (up to 6 months) storage is liquid at 2-8 °C. For longer term storage, aliquot and freeze.
Candida albicans is the most frequently isolated fungal pathogen of humans, affecting immuocompromised patients ranging from premature infants to AIDS sufferers. Systemic infections have an attributed mortality of 30-50%. C. albicans is a diploid organism which has eight sets of homologous chromosomes. Its genome size is about 16 Mb (haploid), about 30% greater than S. cerevisiae (baker ft.s yeast).
Specifications
Specifications
Candida albicans
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Fungi
ELISA, Immunoblot, Immunofluorescence, Immunohistochemistry
4-5 mg/mL
PBS with 0.1% sodium azide; pH 7.2
C. albicans
Candida albicans, type A.
Purified
RUO
Antibody
Polyclonal