Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Canine Heartworm, HRP, Polyclonal, Invitrogen™

Catalog No. PIPA128353
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA128353 100 μL
1 options

Catalog No. PIPA128353

Supplier: Thermo Scientific PA128353

Goat Polyclonal Antibody

Canine heartworm is a common parasite of dogs worldwide. Heartworm occurs in about 30 other species as well but rarely affects humans. The infection is commonly transmitted by mosquitoes. Adult heartworms can grow to be up to 10 inches in length and can live for 5-7 years in dogs affecting the heart and lungs.

PA1-28353 detects Canine Heartworm from nematode samples.PA1-28353 has been successfully used in immunohistochemistry, Western blot applications.The PA1-28353 immunogen is: Dirofiliaria immitis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Canine Heartworm/Dirofilaria immitis
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1.5mg/mL
Conjugate HRP
Gene Alias Adult canine heartworm, Filaria, dirofilariasis
Host Species Goat
Immunogen Dirofiliaria immitis.
Purification Method IgG fraction
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Nematode
Content And Storage Store at 2°-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.