Learn More
IgG (H+L) Sheep anti-Canine, HRP, Invitrogen™
Sheep Polyclonal Secondary Antibody
Manufacturer:  Invitrogen PA184644
Catalog No. PA184644
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Thermo Scientific Anti-Canine secondary antibodies are affinity-purified antibodies with well-characterized specificity for canine (dog) immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Specifications
Specifications
Canine IgG (H+L)
Polyclonal
HRP
Liquid
Purified canine IgG.
Affinity chromatography
RUO
Secondary Antibody
Polyclonal
ELISA
1 mg/mL
PBS with 0.2% BSA and 0.1% Proclin 300
Sheep
IgG
1 mg
4° C, do not freeze
Secondary
Canine