CAPG, Mouse, Clone: 1F1, Abnova™

Mouse monoclonal antibody raised against a full-length recombinant CAPG.

Manufacturer: Abnova Corporation H00000822M01

Catalog No. 89-016-423

  Check Availability
Add to Cart

    Specifications

    Formulation In 1X PBS, pH 7.4
    Gene CAPG
    Gene Accession No. BC000728
    Gene Alias AFCP, MCP
    Gene Symbols CAPG
    Applications Enzyme Linked Immunosorbent Assay,Immunofluorescence,Sandwich ELISA,Western Blotting
    Regulatory Status RUO
    Species Reactivity Human
    Storage Requirements Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
    Gene ID (Entrez) 822
    Description capping protein (actin filament), gelsolin-like
    Antigen CAPG
    Format Liquid
    Immunogen CAPG (AAH00728, 1 a.a. ∽349 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26kDa
    Quantity 100μg
    Primary or Secondary Primary
    Monoclonal or Polyclonal Monoclonal
    Clone 1F1
    Conjugate Unlabeled
    Cross Reactivity Human
    Host Species Mouse
    Isotype IgG1, κ

    This gene encodes a member of the gelsolin/villin family of actin-regulatory proteins. The encoded protein reversibly blocks the barbed ends of F-actin filaments in a Ca2+ and phosphoinositide-regulated manner, but does not sever preformed actin filaments. By capping the barbed ends of actin filaments, the encoded protein contributes to the control of actin-based motility in nonmuscle cells. Alternatively spliced transcript variants have been observed, but have not been fully described. [provided by RefSeq

    Sequence: MYTAIPQSGSPFPGSVQDPGLHVWRVEKLKPVPVAQENQGVFFSGDSYLVLHNGPEEVSHLHLWIGQQSSRDEQGACAVLAVHLNTLLGERPVQHREVQGNESDLFMSYFPRGLKYQEGGVESAFHKTSTGAPAAIKKLYQVKGKKNIRATERALNWDSFNTGDCFILDLGQNIFAWCGGKSNILERNKARDLALAIRDSERQGKAQVEIVTDGEEPAEMIQVLGPKPALKEGNPEEDLTADKANAQAAALYKVSDATGQMNLTKVADSSPFALELLISDDCFVLDNGLCGKIYIWKGRKANEKERQAALQVAEGFISRMQYAPNTQVEILPQGRESPIFKQFFKDWK

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit