Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Capicua, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NB11059905SS

 View more versions of this product

Catalog No. NB11059905T


Add to cart

Description

Description

Capicua Polyclonal specifically detects Capicua in Human, Mouse, Rat, Canine samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Western Blot (Negative).
Specifications

Specifications

Capicua
1 mg/mL
Immunocytochemistry/Immunofluorescence 1:500, Immunoprecipitation 2-5 ug/mg lysate, Western Blot (Negative)
Q96RK0
capicua homolog (Drosophila), KIAA0306capicua (Drosophila) homolog, protein capicua homolog
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat, Canine
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
Affinity Purified
CIC
The epitope recognized by this antibody maps to a region within residue 1150-1200 of human Capicua. [NP_055940.3]
164 kDa
0.025 mg
Cancer
Primary
23152
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only