Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

calpain 5, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human CAPN5 protein.

Manufacturer:  Abnova Corporation H00000726B01

Catalog No. 89-005-005


Add to cart

Description

Description

Calpains are calcium-dependent cysteine proteases involved in signal transduction in a variety of cellular processes. A functional calpain protein consists of an invariant small subunit and 1 of a family of large subunits. CAPN5 is one of the large subunits. Unlike some of the calpains, CAPN5 and CAPN6 lack a calmodulin-like domain IV. Because of the significant similarity to Caenorhabditis elegans sex determination gene tra-3, CAPN5 is also called as HTRA3. [provided by RefSeq

Sequence: MFSCVKPYEDQNYSALRRDCRRRKVLFEDPLFPATDDSLYYKGTPGPAVRWKRPKGICEDPRLFVDGISSHDLHQGQVGNCWFVAACSSLASRESLWQKVIPDWKEQEWDPEKPNAYAGIFHFHFWRFGEWVDVVIDDRLPTVNNQLIYCHSNSRNEFWCALVEKAYAKLAGCYQALDGGNTADALVDFTGGVSEPIDLTEGDFANDETKRNQLFERMLKVHSRGGLISASIKAVTAADMEARLACGLVKGHAYAVTDVRKVRLGHGLLAFFKSEKLDMIRLRNPWGEREWNGPWSDTSEEWQKVSKSEREKMGVTVQDDGEFWMTFEDVCRYFTDIIKCRVINTSHLSIHKTWEEARLHGAWTLHEDPRQNRGGGCINHKDTFFQNPQYIFEVKKPEDEVLICIQQRPKRSTRREGKGENLAIGFDIYKVEENRQYRMHSLQHKAASSIYINSRSVFLRTDQPEGRYVIIPTTFEPGHTGEFLLRVFTDVPSNCRELRLDEPPHTCWSSLCGYPQLVTQVHVLGAAGLKDSPTGANSYVIIKCEGDKVRSAVQKGTSTPEYNVKGIFYRKKLSQPITVQVWNHRVLKDEFLGQVHLKADPDNLQALHTLHLRDRNSRQPSNLPGTVAVHILSSTSLMAV
Specifications

Specifications

calpain 5
Unconjugated
No additive
NM_004055.4
FLJ46245|HTRA3|nCL-3
Mouse
50 uL
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
726
Western Blot
Mouse polyclonal antibody raised against a full-length human CAPN5 protein.
CAPN5
Serum
CAPN5
CAPN5 (NP_004046.2, 1 a.a. ~ 640 a.a) full-length human protein.
RUO
Y
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit