Learn More
anti-CAPS2, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus Biologicals NBP176898
Catalog No. NBP176898
Add to cart
Description
Description
CAPS2 Polyclonal antibody specifically detects CAPS2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
Specifications
CAPS2
1 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 20 ug/ml, Immunohistochemistry-Paraffin 1:10-1:500
NP_060424
calcyphosin-2, calcyphosine 2, Calcyphosine-2, calcyphosphine 2, FLJ34520
Rabbit
IgG
Peptide affinity purified
RUO
Primary
84698
Human, Mouse, Rat
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS with 0.02% Sodium Azide
Affinity Purified
CAPS2
Antibody was raised against a 19 amino acid synthetic peptide near the center of the human CAPS2. The immunogen is located within amino acids 510 - 560 of CAPS2.
150 kDa
0.1 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Multiple isoforms of CAPS2 are known to exist. This CAPS2 antibody is predicted to be specific to CAPS2 and not recognize CAPS1.