Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPZB Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA521372

Catalog No. PIPA521372


Add to cart

Description

Description

PA5-21372 targets CapZ-beta in IHC (P) and WB applications and shows reactivity with Human and mouse samples. The PA5-21372 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 1 and 256 () of CAPZB. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

CAPZB
Polyclonal
Unconjugated
CAPZB
Liquid
CAPZB
Recombit protein fragment contain a sequence corresponding to a region within amino acids 1 and 256 () of CAPZB
Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
2 mg/mL
PBS with 40% glycerol and 0.01% thimerosal; pH 7
P47756, P47757
RP4-657E11.7, CAPB, CAPPB, CAPZ, F-actin-capping protein subunit beta, capZ beta
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
12345, 832
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit