Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAPZA1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA514399

Catalog No. PIPA514399

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

CAPZA1 is a member of the F-actin capping protein alpha subunit family. The protein regulates growth of the actin filament by capping the barbed end of growing actin filaments.
Specifications

Specifications

CAPZA1
Polyclonal
PBS with 0.09% sodium azide
P52907
F-actin-capping protein subunit alpha-1; CapZ alpha-1; CAPZA1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
829
Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CAPZA1
Liquid
CAPZA1
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human CAPZA1
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit