Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carabin Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA520394

Catalog No. PIPA520394


Add to cart

Description

Description

A suggested positive control is Daudi cell lysate. PA5-20394 can be used with blocking peptide PEP-0511.

Antigen binding by the T-cell receptor (TCR) is one of the critical first steps in the immune response, triggering a cascade of signaling pathways that ultimately lead to T-cell activation. Screening a yeast two-hybrid screen of a human T-cell cDNA library with calcineurin, a protein phosphatase involved in multiple signaling pathways including T-cell activation, resulted in the identification of Carabin, a member of the TBC1 domain family of proteins, as a calcineurin-binding protein. Unlike other members of the TBC1 domain protein family which are thought to have a role in regulating cell growth and differentiation, further experiments demonstrated that Carabin is part of a negative regulatory loop for the intracellular TCR signaling pathway as well as an inhibitor of the Ras signaling pathway, suggesting that Carabin may also mediate crosstalk between calcineurin and Ras. Carabin antibody does not recognize TBC1D10A or TBC1D10B. Carabin is known to exist in multiple isoforms.
Specifications

Specifications

Carabin
Polyclonal
Unconjugated
TBC1D10C
Liquid
TBC1D10C
A 16 amino acid peptide from near the carboxy terminus of human Carabin.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q8IV04, Q8C9V1
CARABIN, EPI64C; carabin
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
108995, 374403
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit