Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Carbohydrate Sulfotransferase 5/CHST5, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP136947

Catalog No. NBP136947


Add to cart

Description

Description

Carbohydrate Sulfotransferase 5/CHST5 Polyclonal antibody specifically detects Carbohydrate Sulfotransferase 5/CHST5 in Mouse, Rat samples. It is validated for Peptide ELISA.
Specifications

Specifications

Carbohydrate Sulfotransferase 5/CHST5
0.5 mg/mL
Peptide ELISA Detection limit 1:32000
NP_064334.1
carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 5, gn6st-3, I-GlcNAc6ST
Goat
IgG
0.1 mg
Vision
Primary
23563
ELISA
Unconjugated
0.5 mg/ml Tris (pH 7.3) and 0.5% BSA with 0.02% Sodium Azide
Affinity Purified
CHST5
Peptide with sequence C-DSFKWASSTEKQPE corresponding to C-Terminus according to Uniprot Mouse NP_064334.1.
Immunogen affinity purified
RUO
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only