Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carbonic Anhydrase III Sheep anti-Human, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA175061

Catalog No. PIPA175061


Add to cart

Description

Description

PA1-75061 specifically detects Carbonic Anhydrase 3. PA1-75061 has been successfully used in ELISA applications. The PA1-75061 immunogen is Recombit CAIII.

Carbonic anhydrase III (CAIII) is a member of a multigene family (at least six separate genes are known) that encodes carbonic anhydrase isozymes. These carbonic anhydrases are a class of metalloenzymes that catalyze the reversible hydration of carbon dioxide and are differentially expressed in a number of cell types. The expression of the CA3 gene is strictly tissue specific and present at high levels in skeletal muscle and much lower levels in cardiac and smooth muscle. A proportion of carriers of Duchenne muscle dystrophy have a higher CA3 level than normal. The gene spans 10.3 kb and contains seven exons and six introns.
Specifications

Specifications

Carbonic Anhydrase III
Polyclonal
PBS with 0.09% sodium azide
P07451
CAIII (Muscle)
Sheep
IgG
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
761
ELISA
Unconjugated
CA3
Liquid
CA3
Recombit human CAIII
Affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit