Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carbonic Anhydrase IX/CA9 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
$277.00 - $554.50
Specifications
Antigen | Carbonic Anhydrase IX/CA9 |
---|---|
Concentration | 1.0 mg/mL |
Dilution | Western Blot 1 - 3 ug/ml, Simple Western 1:50, Flow Cytometry 1:1000, ELISA reported in scientific literature (PMID 19963243), Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2 - 5 ug/ml, Immunoprecipitation 1:10 - 1:500. Use reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin 1:200 - 1:500, Immunohistochemistry-Frozen 1:200 - 1:500, Immunoblotting, Gel Super Shift Assays, Proximity Ligation Assay, Microarray reported in scientific literature (PMID 31955345), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Dual RNAscope ISH-IHC 1:1000 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), In situ PLA |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB100417SS
|
Novus Biologicals
NB100417SS |
0.025 mL |
Each for $277.00
|
|
|||||
NB100417
|
Novus Biologicals
NB100417 |
0.1 mL |
Each for $554.50
|
|
|||||
Description
Carbonic Anhydrase IX/CA9 Polyclonal specifically detects Carbonic Anhydrase IX/CA9 in Human, Mouse, Rat, Plant samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Gel Super Shift Assays, Proximity Ligation Assay, Microarray, Dual RNAscope ISH-IHC.Specifications
Carbonic Anhydrase IX/CA9 | |
Western Blot 1 - 3 ug/ml, Simple Western 1:50, Flow Cytometry 1:1000, ELISA reported in scientific literature (PMID 19963243), Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2 - 5 ug/ml, Immunoprecipitation 1:10 - 1:500. Use reported in multiple pieces of scientific literature, Immunohistochemistry-Paraffin 1:200 - 1:500, Immunohistochemistry-Frozen 1:200 - 1:500, Immunoblotting, Gel Super Shift Assays, Proximity Ligation Assay, Microarray reported in scientific literature (PMID 31955345), Chromatin Immunoprecipitation (ChIP) 1:10-1:500, Dual RNAscope ISH-IHC 1:1000 | |
Polyclonal | |
Rabbit | |
Cancer, Cellular Markers, HIF Target Genes, Hypoxia, Lipid and Metabolism, Signal Transduction | |
PBS with 0.02% Sodium Azide | |
CA-IX, CAIXcarbonic anhydrase 9, Carbonate dehydratase IX, carbonic anhydrase IXpMW1, carbonic dehydratase, EC 4.2.1.1, G250, Membrane antigen MN, MNRCC-associated antigen G250, P54/58N, RCC-associated protein G250, Renal cell carcinoma-associated antigen G250 | |
CA9 | |
IgG | |
Affinity Purified | |
Human, dog, rat and mouse (PMID 22842475). |
1.0 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunoblot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), In Situ Hybridization (ISH), In situ PLA | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Canine, Plant | |
Q16790 | |
768 | |
A synthetic peptide made to a C-terminal sequence of human Carbonic Anhydrase IX (within residues 400-459) [UniProt Q16790] | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. | |
55 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title