Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CES1 Monoclonal Antibody (544623), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA524244
Description
In direct ELISAs and Western blots, 100% cross-reactivity with recombinant mouse (rm) CES-3 and no cross-reactivity with rmCES-2, rmCES-5, or recombinant human CES-2 is observed. Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
CES1 Monoclonal antibody specifically detects CES1 in Human samples. It is validated for Immunoprecipitation, Western Blot.Specifications
CES1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P23141 | |
CES1 | |
Mouse myeloma cell line NS0-derived recombinant human Carboxylesterase1/CES-1 His19-Glu563 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Immunoprecipitation, Western Blot | |
544623 | |
Unconjugated | |
CES1 | |
ACAT; acyl coenzyme A:cholesterol acyltransferase; acyl-coenzyme A:cholesterol acyltransferase; Brain carboxylesterase hBr1; carboxyesterase ES-3; carboxylesterase 1; carboxylesterase 1 (monocyte/macrophage serine esterase 1); carboxylesterase 1A; carboxylesterase 1E; Carboxylesterase 1G; carboxylesterase 1-like; carboxylesterase 2 (liver); CE-1; CEH; Ces1; Ces-1; Ces1a; Ces1e; Ces1g; Ces1l; CES2; cholesteryl ester hydrolase; cocaine carboxylesterase; Eg; EG244595; Egasyn; Es22; Es-22; ES-HTEL; esterase 22; esterase-22; ES-x; Gm4976; hCE-1; HMSE; HMSE1; human monocyte/macrophage serine esterase 1; liver carboxylesterase 1; Liver carboxylesterase 22; liver carboxylesterase 3; methylumbelliferyl-acetate deacetylase 1; MGC117365; Monocyte/macrophage serine esterase; PCE-1; pI 5.5 esterase; REH; Retinyl ester hydrolase; serine esterase 1; SES1; Ses-1; TGH; triacylglycero; Triacylglycerol hydrolase | |
Mouse | |
Protein A/G | |
RUO | |
1066 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction