Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Carboxypeptidase B1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA528204

Catalog No. PIPA528204


Add to cart

Description

Description

PA5-28204 targets Carboxypeptidase B in WB applications and shows reactivity with Human samples. The PA5-28204 immunogen is recombit fragment corresponding to a region within amino acids 1 and 259 of Human Carboxypeptidase B. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Three different procarboxypeptidases A and two different procarboxypeptidases B have been isolated. The B1 and B2 forms differ from each other mainly in isoelectric point. Carboxypeptidase B1 is a highly tissue-specific protein and is a useful serum marker for acute pancreatitis and dysfunction of pancreatic transplants. It is not elevated in pancreatic carcinoma.
Specifications

Specifications

Carboxypeptidase B1
Polyclonal
Unconjugated
CPB1
Liquid
CPB1
Recombit fragment corresponding to a region within amino acids 1 and 259 of Human Carboxypeptidase B
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P15086
CPB, PASP, PCPB, carboxypeptidase B, pancreas-specific protein, pancreatic carboxypeptidase B, procarboxypeptidase B, protaminase, tissue carboxypeptidase B
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1360
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit