Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Carboxypeptidase M Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521727
Description
Recommended positive controls: HepG2, Rat liver. Predicted reactivity: Mouse (87%), Rat (86%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Carboxypeptidase M Polyclonal antibody specifically detects Carboxypeptidase M in Human, Rat samples. It is validated for Western Blot.Specifications
Carboxypeptidase M | |
Polyclonal | |
Unconjugated | |
CPM | |
1110060I01Rik; 5730456K23Rik; AA589379; Carboxypeptidase M; Cpm; E030045M14Rik; renal carboxypeptidase; urinary carboxypeptidase B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1368, 314855 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
P14384 | |
CPM | |
Recombinant fragment corresponding to a region within amino acids 43 and 303 of Carboxypeptidase M (Uniprot ID#P14384) | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction