Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cardiac Troponin I Mouse, Alexa Fluor 647, Clone: C5, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564409

Catalog No. BDB564409


Add to cart

Description

Description

The C5 monoclonal antibody specifically binds to Cardiac Troponin I (cTnI). cTnI is a ~30-kDa protein that is also known as TNNI3 and TNNC1. Troponin I is the inhibitory subunit of Troponin, which also contains the subunits Troponin C and Troponin T. Troponin aids the Ca[2+]-dependent interaction of actin and myosin, which are the main contracile components in muscle cells. cTnI is specifically expressed in cardiac tissue, while skeletal troponin I is expressed in skeletal muscle in slow and fast isoforms. Hereditary cardiomyopathy is associated with mutations in cTnI. During product development, the purified C5 monoclonal antibody detected cTnI by western blot analysis of cellular lysates and flow cytometric analysis of fixed and permeabilized cells.

Specifications

Specifications

Cardiac Troponin I
C5
cTnI, TNNI3, TNNC1
Affinity Purified
Human
50 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
Alexa Fluor 647
Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide.
Murine
IgG2b, κ
RUO
Primary
Human, Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit