Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cardiac Troponin I Mouse anti-Human, Clone: 3C7, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA120115
Description
MA1-20115 detects cardiac troponin 1 in human samples. MA1-20115 has been successfully used in ELISA and Western blot procedures. The MA1-20115 immunogen is full length, human cardiac troponin 1 protein.
Cardiac Troponin I (Cardiac-specific troponin I, TnI, Troponin I) is an integral inhibitory protein in cardiac muscle that exists as part of a complex with troponin C (TnC) and troponin T (TnT). The TnT subunit of troponin binds to tropomyosin-t form, a troponin-tropomyosin complex, anchored in place by the binding of TnI to actin, within muscle thin filaments. Structural change resulting from the binding of calcium to specific sites on the regulatory TnC subunit, releases the inhibitory region of TnI from actin, anabling the attachment of the molecular motor protein myosin, allowing for muscle contraction and hence movement. The measurements of blood TnI and TnT levels is an important diagnostics indicator of heart muscle damage, and can be used to differentiate between angina and myocardial infarction in patients with chest pains. Cardiac Troponin I has also been shown to inhibit angiogenesis in vivo and in vitro. Mutations in the Cardiac Troponin I gene cause familial hypertrophic cardiomyopathy type 7 (CMH7) and familial restrictive cardiomyopathy (RCM).Specifications
Cardiac Troponin I | |
Monoclonal | |
Unconjugated | |
P19429 | |
TNNI3 | |
Full length, human cardiac troponin 1 protein. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Western Blot | |
3C7 | |
TNNI3 | |
CMD2A, CMH7, MGC116817, RCM1, TNNC1, cTnI | |
Mouse | |
Protein A | |
RUO | |
7137 | |
4° C | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title