Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cardiotrophin 1 Monoclonal Antibody (89215), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA523846
Description
Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.
Cardiotrophin 1 Monoclonal antibody specifically detects Cardiotrophin 1 in Human samples. It is validated for Neutralization, Western Blot.Specifications
Cardiotrophin 1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q16619 | |
CTF1 | |
E. coli-derived recombinant human Cardiotrophin-1/CT-1 Ser2-Ala201 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Neutralization, Western Blot | |
89215 | |
Unconjugated | |
CTF1 | |
cardiophin 1; Cardiotrophin; cardiotrophin 1; Cardiotrophin1; cardiotrophin-1; CT 1; CT1; CT-1; CT-1CT1; CTF 1; Ctf1; IL-6 cytokine family | |
Mouse | |
Protein A/G | |
RUO | |
1489 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction