Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CAS Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA137229

Catalog No. 01-673-154


Add to cart

Description

Description

This antibody is predicted to react with bovine, amphibian, mouse based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is liver carcinoma.

CAS (cellular apoptosis susceptibility contains 971 amino acids with 59% overall sequence homology as compared to yeast chromosome segregation homolog, CSE1. CAS is highly expressed in testis and fetal liver. CAS expression in human fibroblasts increases as cells are induced to proliferate and decreases in growth arrested cells.
Specifications

Specifications

CAS
Polyclonal
PBS with 1% BSA and <0.1% sodium azide
Q9ERK4, P55060
CAS, CSE1, XPO2, cellular apoptosis susceptibility protein, chromosome segregation 1-like protein, exp2, exportin-2, importin-alpha re-exporter
Rabbit
IgG
1 mL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
110750, 1434
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CSE1L
Liquid
CSE1L
Synthetic peptide from N-terminus of human CAS.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit