Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CSN1S1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513490

Catalog No. PIPA513490

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CASA play an important role in the capacity of milk to transport calcium phosphate. Casoxin D acts as opioid antagonist and has vasorelaxing activity mediated by bradykinin B1 receptors.
Specifications

Specifications

CSN1S1
Polyclonal
PBS with 0.09% sodium azide
P47710
Alpha-S1-casein; Casoxin-D; CSN1; CASA; CSN1S1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1446
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
CSN1S1
Liquid
CSN1S1
KLH conjugated synthetic peptide between 42-70 amino acids from the central region of human CASA
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit