Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASC4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524013

Catalog No. PIPA524013


Add to cart

Description

Description

This antibody is predicted to react with mouse based on sequence homology.

The increased expression level of this gene is associated with HER-2/neu proto-oncogene overexpression. Amplification and resulting overexpression of this proto-oncogene are found in approximately 30% of human breast and 20% of human ovarian cancers. Further characterization of the product of this gene may yield insight into the fundamental biology and pathogenetic effects of HER-2/neu overexpression in human breast and ovarian cancer cells. Alternatively spliced variants encoding different isoforms have been identified for this gene.
Specifications

Specifications

CASC4
Polyclonal
PBS with 0.09% sodium azide
Q6P4E1, Q6P2L7
H63, cancer susceptibility candidate gene 4 protein, gene associated with HER-2/neu overexpression, protein CASC4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
113201, 319996
Flow Cytometry, Western Blot
Unconjugated
CASC4
Liquid
CASC4
KLH conjugated synthetic peptide between 336-365 amino acids from the C-terminal region of human CASC4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit