Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CK1 alpha Chicken anti-Bovine, Chicken, Equine, Human, Mouse, Porcine, Rat, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA110006

Catalog No. 01-670-263


Add to cart

Description

Description

Casein kinases are operationally defined by their preferential utilization of acidic proteins such as caseins as substrates. CK1a can phosphorylate a large number of proteins. This cytoplasmic protein participates in Wnt signaling. It has been demonstrated to phosphorylate CTNNB1 on Ser-45 and to interact with the Axin complex.
Specifications

Specifications

CK1 alpha
Polyclonal
PBS with 0.02% sodium azide
P67827, P67962, P48729, Q8BK63, O19175, P97633
YIL035C
Chicken
IgY
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100060556, 100514516, 113927, 1452, 282684, 395924, 93687
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Unconjugated
CSNK1A1
Liquid
CSNK1A1
Recombit full length CK1a (shortest isoform)
PEG precipitation
RUO
Antibody
Polyclonal
Bovine, Chicken, Equine, Human, Mouse, Porcine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit