Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CK1 delta Goat anti-Human, Polyclonal, Invitrogen™

Goat Polyclonal Antibody

Manufacturer:  Invitrogen PA128034

Catalog No. PIPA128034


Add to cart

Description

Description

PA1-28034 detects Casein Kinase 1 delta from human samples. PA1-28034 has been successfully used in immunohistochemistry (paraffin tissue) applications. The PA1-28034 immunogen is: Synthetic peptide, corresponding to amino acids 310 - 327 of Human Casein Kinase 1 delta.

This gene is a member of the casein kinase I (CKI) gene family whose members have been implicated in the control of cytoplasmic and nuclear processes, including DNA replication and repair. The encoded protein is highly similar to the mouse and rat CK1 delta homologs. Two transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

CK1 delta
Polyclonal
whole serum with 0.1% sodium azide
P48730
HCKID
Goat
IgG
RUO
Antibody
Polyclonal
Human
ELISA, Immunohistochemistry (Paraffin)
Unconjugated
CSNK1D
Liquid
CSNK1D
Synthetic peptide, corresponding to amino acids 310 - 327 of Human Casein Kinase 1 delta.
50 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1453
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit