Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CK2 beta Mouse anti-Human, Mouse, Rat, Clone: 26C3, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific LFMA0216
Description
A suggested positive control for this product is K562 cells.
This gene encodes the beta subunit of casein kinase II, a ubiquitous protein kinase which regulates metabolic pathways, signal transduction, transcription, translation, and replication. The enzyme is composed of three subunits, alpha, alpha prime and beta, which form a tetrameric holoenzyme. The alpha and alpha prime subunits are catalytic, while the beta subunit serves regulatory functions. The enzyme localizes to the endoplasmic reticulum and the Golgi apparatus.Specifications
CK2 beta | |
Monoclonal | |
Unconjugated | |
P67870, P67871, P67874 | |
CSNK2B | |
Recombit human protein purified from E.coli (His-casein kinase II beta) | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 λ |
ELISA, Western Blot | |
26C3 | |
CSNK2B | |
DADB-127H9.2, CK2B, CK2N, CSK2B, G5A, MGC138222, MGC138224 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
13001, 1460, 81650 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title