Learn More
Phospho-CK2 beta (Ser209), Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA128095
Description
This gene encodes the beta subunit of casein kinase II, a ubiquitous protein kinase which regulates metabolic pathways, signal transduction, transcription, translation, and replication. The enzyme is composed of three subunits, alpha, alpha prime and beta, which form a tetrameric holoenzyme. The alpha and alpha prime subunits are catalytic, while the beta subunit serves regulatory functions. The enzyme localizes to the endoplasmic reticulum and the Golgi apparatus.
PA1-28095 detects Casein Kinase 2 beta from human samples.PA1-28095 has been successfully used in Western blot applications.The PA1-28095 immunogen is: Synthetic phosphopeptide (Human) from the region of casein kinase 2 that contains serine 209.Specifications
Phospho-Casein kinase II beta pSer209 | |
Polyclonal | |
Unconjugated | |
P67870 | |
CSNK2B | |
Synthetic phosphopeptide (Human) from the region of casein kinase 2 that contains serine 209. | |
50 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
0.25mg/mL | |
CSNK2B | |
Ck2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1460 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Purified |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.