Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CASKIN2, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP152628

Catalog No. NBP152628


Add to cart

Description

Description

CASKIN2 Polyclonal antibody specifically detects CASKIN2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

CASKIN2
1.0 mg/mL
Western Blot, Immunoprecipitation 2-5 mcg/mg
Affinity Purified
CASKIN2
The immunogen for this product maps to a region between residue 700 to 750 of human CASK Interacting Protein 2 using the numbering given in entry NP_065804.2 (GeneID 57513).
Immunogen affinity purified
RUO
Primary
57513
Immunoprecipitation, Western Blot
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
ANKS5B, CASK interacting protein 2, caskin-2, FLJ21609, KIAA1139
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only