Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase 3 (Cleaved Asp175) Rabbit anti-Human, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523921

Catalog No. PIPA523921

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a protein which is a member of the cysteine-aspartic acid protease (caspase) family. Sequential activation of caspases plays a central role in the execution-phase of cell apoptosis. Caspases exist as inactive proenzymes which undergo proteolytic processing at conserved aspartic residues to produce two subunits, large and small, that dimerize to form the active enzyme. This protein cleaves and activates caspases 6, 7 and 9, and the protein itself is processed by caspases 8, 9 and 10. It is the predomit caspase involved in the cleavage of amyloid-beta 4A precursor protein, which is associated with neuronal death in Alzheimer ft.s disease. Alternative splicing of this gene results in two transcript variants that encode the same protein.
Specifications

Specifications

Caspase 3 (Cleaved Asp175)
Polyclonal
Unconjugated
CASP3
Liquid
CASP3
KLH conjugated synthetic peptide between 149-179 amino acids from human CASP3(Asp175)
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Rat
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
P42574
CPP32, CPP32B, SCA-1, CASP-3, CPP-32, PARP cleavage protease, SREBP cleavage activity 1, Yama, apopain, caspase 3, apoptosis-related cysteine protease, caspase-3, cysteine protease CPP32, procaspase3, protein Yama
Rabbit
IgG
400 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
836
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit