Learn More
Caspase 1 Rabbit anti-Human, Polyclonal, MilliporeSigma™ Available on GSA/VA Contract for Federal Government customers only.
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 06503I
Catalog No. 06-503-IMI
Add to cart
Description
Description
Anti-Caspase 1 Antibody is a highly specific rabbit polyclonal antibody, that targets Caspase 1 and has been tested in western blotting and immunohistochemistry (Paraffin).
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Caspase 1
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
CASP1; IL1BC; IL1BCE
KLH-conjugated linear peptide corresponding to the p20 subunit of human Caspase 1.
100μg
Primary
NP_001214