Learn More
Caspase 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA126382
Catalog No. PA126382
Add to cart
Description
Description
PA1-26382 detects Caspase 1 from human, rat, and mouse samples. PA1-26382 has been successfully used in immunohistochemistry (paraffin) procedures. Formalin fixed paraffin embedded tissue sections require high temperature antigen unmasking with all 10 mM citrate buffer, pH 6.0 prior to immunostaining. The PA1-26382 immunogen is a synthetic peptide corresponding to the second quarter of human Caspase 1 protein.
Caspases are a family of cysteine proteases that are key mediators of programmed cell death or apoptosis. The precursor form of all caspases is composed of a prodomain, and large and small catalytic subunits. The active forms of caspases are generated by several stimuli including ligand-receptor interactions, growth factor deprivation and inhibitors of cellular functions.
Specifications
Specifications
Caspase 1
Polyclonal
PBS with sodium azide
P29466, P29452, P43527
ICE, Il1bc
Rabbit
IgG
500 μL
4° C
Primary
12362, 25166, 834
Immunohistochemistry (Paraffin)
Unconjugated
CASP1
Liquid
CASP1
Synthetic peptide corresponding to the second quarter of human Caspase 1 protein.
Protein A
RUO
Antibody
Polyclonal
Human, Mouse, Rat