Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase 10 Rabbit anti-Chicken, Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14300

Catalog No. PA14300


Add to cart

Description

Description

PA1-4300 detects the Caspase 10 protein in human, mouse, rat, and chicken samples. PA1-4300 has been successfully used in Western blot procedures. PA1-4300 immunizing peptide is the N terminal sequence of all isoforms of human Caspase-10.

Caspases are a family of cysteine proteases that are key mediators of programmed cell death or apoptosis. The precursor form of all caspases is composed of a prodomain, and large and small catalytic subunits. The active forms of caspases are generated by several stimuli including ligand-receptor interactions, growth factor deprivation and inhibitors of cellular functions.
Specifications

Specifications

Caspase 10
Polyclonal
Unconjugated
CASP10
Liquid
CASP10
Synthetic peptide to the N terminus: M(1) K S Q G Q H W Y S S S D K N(15)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Chicken, Human, Mouse, Rat
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q92851
FLICE2, Mch4, CASP-10, ICE-like apoptotic protease 4, Apoptotic protease Mch-4, FAS-associated death domain protein interleukin-1B-converting enzyme 2
Rabbit
IgG
100 μg
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Primary
424081, 843
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit