Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-11 Rat anti-Mouse, Unlabeled, Clone: 17D9, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 564971

Catalog No. BDB564971


Add to cart

Description

Description

The 17D9 monoclonal antibody specifically binds to mouse Caspase-11 (CASP-11/Casp11) which is also known as ICH-3. Mouse Caspase-11 is homologous to human Caspase-4 and is encoded by the mouse Casp4 (Caspase 4, apoptosis-related cysteine peptidase) gene. Caspase-11 is a cysteine protease that functions in apoptosis and inflammatory responses. Caspase-11 exists as 38 and 43 kDa pro-caspase forms, and as 30 and 10 kDa active forms. Caspase-11 is an initiator caspase that activates other caspases, including Caspase-1 [ IL-1β -converting enzyme (ICE)], Caspase-3, and Caspase-7, which are involved in executing apoptosis. When Caspase-11 is deficient, cells are unable to process or secrete Interleukin-1 β ( IL-1β ) and demonstrate defective motility in response to certain stimuli. Unlike other caspases that are expressed constitutively, Caspase-11 expression can be upregulated in cells stimulated with certain activators, e.g., macrophages, lymphocytes and hepatocytes stimulated with lipopolysaccharide.

Specifications

Specifications

Caspase-11
17D9
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
100μg
Store undiluted at 4°C
Monoclonal
Western Blot
0.5mg/mL
CASP-11; Casp11; CASP-4; Casp4; Caspase-4; Caspl; ich-3; Ich3
Affinity Purified
Mouse Caspase-11 Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit