Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase 12 Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA130776

Catalog No. PIPA130776


Add to cart

Description

Description

PA1-30776 detects Caspase 12 from human, mouse, rat samples. PA1-30776 has been successfully used in Western blot and IHC (P) applications. The PA1-30776 immunogen is: Synthetic peptide - amino acids 100-116 of Mouse Caspase 12 Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Caspases are a family of cysteine proteases that are key mediators of programmed cell death or apoptosis. The precursor form of all caspases is composed of a prodomain, and large and small catalytic subunits. The active forms of caspases are generated by
Specifications

Specifications

Caspase 12
Polyclonal
Unconjugated
CASP12
Liquid
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
100506742, 12364, 156117
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.02% sodium azide
Q6UXS9, O08736, Q920D5
CASP12
Synthetic peptide corresponding to amino acids 100-116 of Mouse Caspase 12
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit