Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase 12 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523369

Catalog No. PIPA523369


Add to cart

Description

Description

Caspases are cysteine proteases that cleave C-terminal aspartic acid residues on their substrate molecules. Caspase-12 (CASP12) process inflammatory cytokines. It attenuates the macrophage-elicited T helper cell type 1 and type 2 cytokine response, with probable compensatory enabling of T cell-derived interferon-gamma formation. Caspase-12 dampened the production of the proinflammatory cytokines interleukin 1-beta, IL-18 and IFNg, but not tumor necrosis factor-alpha and IL-6, in response to various bacterial components that stimulate Toll-like receptor and NOD pathways. In rodents, Casp12 mediates apoptosis in response to endoplasmic reticulum stress. However, in human, Caspase-12 appears to be nonfunctional. A SNP in Caspase-12 in humans results in the synthesis of either a truncated protein (Csp12S) or a full-length caspase proenzyme (Csp12L). SNP encoding Csp12L is confined to populations of African descent and confers hyporesponsiveness to lipopolysaccharide-stimulated cytokine production in ex vivo whole blood but has no significant effect on apoptotic sensitivity. PCR analysis detected highest expression of Caspase-12 in lung. Expression was intermediate in small intestine, stomach, and kidney. Nine Caspase-12 splice variants are identified in lung and small intestine.
Specifications

Specifications

Caspase 12
Polyclonal
Unconjugated
CASP12
Liquid
CASP12
Synthetic peptide (100 GLNIRNKEFNYLHNRNGS 117) of Caspase-12 protein.
Protein A
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
Q6UXS9
UNQ9415, CASP-12, CASP12P1, caspase 12 pseudogene 1, inactive caspase-12
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
100506742
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit