Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-2 Mouse anti-Human, Unlabeled, Clone: 35, BD

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 611022

 View more versions of this product

Catalog No. BDB611022


Add to Cart

Description

Description

Caspase-2/ICH-1 is related to the C. elegans cell death gene product CED-3 and its mammalian homologue interleukin-1β-converting enzyme (ICE). Caspase-2 /ICH-1 was identified from a mouse cDNA library and originally termed NEDD-2. The NEDD-2 mRNA was found to be expressed during early mouse embryonic brain development and subsequently down-regulated in adult neuronal tissue. With the identification of the human NEDD-2 gene, the murine gene was renamed Ich-1 to symbolize Ice and ced-3 homology. Caspase-2/ICH-1 mRNA is alternatively spliced. The larger mRNA species encoding a product of 435 amino acids is known as Caspase-2 long, or ICH-1L. The smaller mRNA species encoding a protein of 312 amino acids is named Caspase-2 short, or ICH-1S. Overexpression of ICH-1L induces apoptosis, while over-expression of Ich-1S suppresses Rat-1 cell death induced by serum deprivation. Thus, it appears that Caspase-2/ICH-1 plays an important dual role in programmed cell death.

Immunofluorescence, Western Blotting

Specifications

Specifications

Caspase-2
Monoclonal
250μg/mL
ICH-1L
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
35
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human ICH-1L aa. 225-401
50 μg
Cell Biology
Human
IgG1
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.