Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Caspase-2, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP177911

Catalog No. NBP177911

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

Caspase-2
0.44 mg/ml
Western Blot 1:750, ELISA 1:1500-1:6000
P55215
caspase-2, CASP-2, caspase 2, apoptosis-related cysteine peptidase, ICH1, NEDD2, NEDD-2, PPP1R57
Rabbit
IgG
0.1 mg
Apoptosis, Cancer, Caspases
Primary
835
Mouse, Rat
ELISA, Western Blot
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Affinity Purified
CASP2
A synthetic peptide corresponding to the N-terminus of rat Caspase-2 protein conjugated to Keyhole Limpet Hemocyanin (KLH) through a cysteine residue linker.
Immunogen affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
This affinity purified antibody is directed against rat and mouse caspase-2. The antibody detects long and short forms of the protein. Antiserum was purified against the immunizing peptide. This antibody reacts with mouse and rat caspase-2, but not caspase-2 from human sources. Cross reactivity with caspase-2 from other sources has not been determined. Rat and mouse caspase-2 show 94% homology. There is no significant homology to the human form.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only