Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caspase-3 Mouse anti-Human, Unlabeled, Clone: 19, BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 610322

 View more versions of this product

Catalog No. BDB610322


Add to cart

Description

Description

Apoptosis, a selective process of genetically programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptotic cells are characterized by loss of cell volume, plasma membrane blebbing, nuclear condensation, chromatin aggregation, and endonucleocytic degradation of DNA into nucleosomal fragments. Caspase-3 (CPP32, Yama, apopain) is a member of the family of cysteine proteases which includes interleukin-1β converting enzyme (ICE) and C. elegans protein, Ced-3. An apoptotic signal such as granzyme B of cytotoxic T-cells (CTLs) or ICE-like proteases induces the intracellular cleavage of Caspase-3 from the inactive pro-form (32kDa) to the active form which consists of the p20, p17, and p12 subunits. The active form of Caspase-3 cleaves several other apoptotic proteins including proteins such as DNA Fragmentation Factor (DFF). Apoptosis can be inhibited by coexpression of Bcl-2 as well as inhibitors of Caspase-3 or other members of the family of cysteine proteases. This antibody recognizes the human pro-form (inactive) of Caspase-3 at 32kDa. In addition, it has been reported to recognize the active form of Caspase-3 at 20-21kDa in apoptotic cell lysates.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Caspase-3
19
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Murine
IgG2a
RUO
Store undiluted at -20°C.
Monoclonal
Western Blot
250μg/mL
CPP32
Affinity Purified
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50μg
Cell Biology
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.