Learn More
MilliporeSigma™ anti-Caspase 3, Clone: 4-1-18,
Manufacturer:  MilliporeSigma™ 05654
Catalog No. 05-654-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
Caspase 3
4-1-18
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
IgG2a
100μg
Primary
Human
Western Blot
Unconjugated
NM_032991
CASP3, apopain, CASP-3, procaspase3, CPP32B, SCA-1, CPP-32, Apopain, CPP32, Yama
N-terminal HIS tagged fusion protein corresponding to full length human Caspase 3
Protein G chromatography
−20°C for 24 months.
Monoclonal