Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CASZ1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523743

Catalog No. PIPA523743

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CASZ1
Polyclonal
PBS with 0.09% sodium azide
Q86V15, Q9CWL2
CST, SRG, ZNF693, dJ734G22.1, castor homolog 1, zinc finger, castor-related protein, putative survival-related protein, survival related, survival-related, zinc finger protein 693, zinc finger protein castor homolog 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
54897, 69743
Western Blot
Unconjugated
CASZ1
Liquid
CASZ1
KLH conjugated synthetic peptide between 1574-1601 amino acids from the C-terminal region of human CASZ1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit