Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Cat IgG, Agarose, Isotype Control, Novus Biologicals™

Feline Isotype Control Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP197131

Catalog No. NBP197131


Add to cart

Description

Description

IgG Isotype Control antibody specifically detects IgG in Feline samples. It is validated for Immunoprecipitation.
Specifications

Specifications

IgG
Agarose
0.01 M Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: None. with 0.01% Sodium Azide
immunoglobulin heavy constant gamma 1 (G1m marker)
Feline
Multi-step
RUO
Isotype Control
3500
Immunoprecipitation
Immunoprecipitation 1:10 - 1:500
Conjugated
IGHG1
IgG
10 mg
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Primary
Feline
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only