Learn More
Seracare Life Sciences IncSupplier Diversity Partner ANTI-CAT IGG (GAMMA) AB HRP  DFS Item
Manufacturer:  Seracare Life Sciences IncSupplier Diversity Partner 52200291
Affinity purified polyclonal secondary antibody against Cat IgG, heavy chains (gamma) made in Goat and labeled with horseradish peroxidase. Product is in lyophilized form.
Catalog No. 50-669-46