Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Cat IgG, Rhodamine, Isotype Control, Novus Biologicals™

Feline Isotype Control Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196754

Catalog No. NBP196754


Add to cart

Description

Description

IgG Isotype Control antibody specifically detects IgG in Feline samples. It is validated for Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

IgG
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide
immunoglobulin heavy constant gamma 1 (G1m marker)
Feline
Ion exchange chromatography
RUO
Isotype Control
3500
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
Rhodamine
Lyophilized
IGHG1
IgG
1 mg
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Primary
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Feline IgG and anti-Feline Serum.
Feline
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only