Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Catalase Rabbit anti-Bovine, Human, Mouse, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523246

Catalog No. PIPA523246


Add to cart

Description

Description

Catalase is a 527 amino acid heme-containing peroxisomal enzyme belonging to the catalase family. It is a common enzyme that occurs in almost all aerobically respiring organisms and activates the decomposition of hydrogen peroxide, thus serving an important role in protecting cells from the toxic effects of hydrogen peroxide. Catalase,  a homotetramer, is associated with cofactors like heme group and NADP and is known to have one of the highest turnover rates of all enzymes. Catalase is a natural antioxidant used to study roles of reactive oxygen species in gene expression and apoptosis. Defects in Catalase lead to a dreadful disease acatalasia, also known as acatalasemia, characterized by the absence of catalase activity in red cells and are often associated with ulcerating oral lesions. Catalase is also known to play an important role in ethanol metabolism, inflammation, apoptosis, aging and cancer.
Specifications

Specifications

Catalase
Polyclonal
Unconjugated
CAT
Liquid
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
12359, 24248, 531682, 847
Immunohistochemistry (Paraffin), Western Blot
1.0 mg/mL
PBS with 0.05% sodium azide
P00432, P04040, P24270, P04762
CAT
Bovine liver catalase.
Protein A
RUO
Antibody
Polyclonal
Bovine, Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit