Learn More
δ-Catenin Mouse, Unlabeled, Clone: 30, BD
Mouse Monoclonal Antibody
$276.00 - $508.00
Specifications
Antigen | δ-Catenin |
---|---|
Clone | 30 |
Concentration | 250μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
The catenins (α, β, γ, δ, p120[ctn]) are cytoplasmic proteins that are related to the Drosophila Armadillo protein. Catenins may have dual roles since they are components of cell-cell adherens junctions and can translocate to the nucleus after stimulation of the Wingless (Wnt-1 homolog) signaling pathway. α-Catenin has two subtypes: αE-Catenin, which is expressed ubiquitously, and αN-Catenin, which is expressed in the nervous system. β-Catenin binds to the cytoplasmic tail of E-Cadherin at adherens junctions and has been implicated in Wnt-1 signaling. γ-Catenin is associated with desmoglein in desmosomes and is closely related to β-Catenin. p120[ctn] is related to both β- and γ-catenin and is a substrate of tyrosine kinases localized at adherens junctions. δ-catenin was identified by its ability to bind the Alzheimer's disease-related protein, presenilin-1. It is most closely related to p120[ctn] and the desmosomal protein, p0071. It contains 10 Armadillo (Arm) repeats, as compared to the 13 Arm repeats found in β-Catenin. δ-Catenin is expressed at high levels in the developing nervous system, where it may be involved in neuronal progenitor cell migration and dendrite development.
Host Species: Mouse
Clone: 30
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse δ-Catenin aa. 85-194
Formula Weight [Chemical]: 160kDa
Immunofluorescence, Western Blotting
Specifications
δ-Catenin | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified |
30 | |
Western Blot | |
Unconjugated | |
RUO | |
Mouse, Rat | |
Mouse δ-Catenin aa. 85-194 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.