Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

delta Catenin Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA139454

Catalog No. PA139454


Add to cart

Description

Description

Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is breast lobular carcinoma.

Catenin delta (p120) is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.
Specifications

Specifications

delta Catenin
Polyclonal
PBS with 1% BSA and 0.1% sodium azide
O60716
CAS, CTNND, P120CAS, P120CTN, p120, p120CAS, p120CTN, cadherin-associated Src substrate, catenin delta-1, p120 catenin
Rabbit
IgG
1 mL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1500
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
CTNND1
Liquid
CTNND1
Synthetic peptide corresponding to C-terminus of human p120.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit