Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Catenin, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA120931

Catalog No. PIPA120931

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Gamma Catenin is a common junctional plaque protein. The membrane-associated plaques are architectural elements in an important strategic position to influence the arrangement and function of both the cytoskeleton and the cells within the tissue. The presence of gamma Catenin in both the desmosomes and in the intermediate junctions suggests that it plays a central role in the structure and function of submembranous plaques.

PA1-20931 detects Gamma Catenin in Human, Mouse and Rat samples.PA1-20931 has been successfully used in Immunohistochemistry (Paraffin) procedures.PA1-20931 immunogen corresponds to Synthetic peptide (Human) (C terminal)..
Specifications

Specifications

gamma Catenin
Polyclonal
PBS with 1% BSA and 0.1% sodium azide; pH 7.4
P14923, Q02257, Q6P0K8
ARVD12, CTNNG, DP3, DPIII, PDGB, PKGB
Rabbit
IgG
7mL
4° C, do not freeze
Primary
3728
Immunohistochemistry (Paraffin)
Unconjugated
JUP
Purified
JUP
Synthetic peptide (Human) (C terminal).
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit