Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-β-Catenin (NT), Clone: E247, Rabbit Monoclonal,
Supplier: MilliporeSigma™ 041002
Specifications
β-Catenin (NT) | |
Monoclonal | |
Unconjugated | |
NP_001091679.1 | |
Synthetic peptide corresponding to residues near the N-terminus of human β-catenin. | |
100 μL | |
Beta-catenin, CTNNB, DKFZp686D02253, FLJ25606, FLJ37923, OTTHUMP00000165222, OTTHUMP00000165223 | |
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles. |
Immunocytochemistry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
E247 | |
Rabbit Monoclonal IgG in buffer containing 50mM Tris-Glycine (pH 7.4), 0.15M NaCl containing 40% Glycerol, 0.01% sodium azide and 0.05% BSA. | |
Rabbit | |
Unpurified | |
Primary | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only