Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Cathepsin B, FITC, Polyclonal, Novus Biologicals™

Sheep Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP125931

Catalog No. NBP125931


Add to cart

Description

Description

Cathepsin B Polyclonal antibody specifically detects Cathepsin B in Human samples. It is validated for ELISA.
Specifications

Specifications

Cathepsin B
1.0mg/mL
ELISA 1:1000-1:10000
P07858
APP secretase, APPS, cathepsin B, Cathepsin B1, CPSBamyloid precursor protein secretase, cysteine protease, EC 3.4.22, EC 3.4.22.1
Sheep
IgG
0.5 ml
Cancer, Cellular Markers, Neuroscience
Primary
1508
Human
ELISA
FITC
PBS with 0.05% Sodium Azide
Purified
CTSB
Native, from placenta
Protein G purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
This antibody detects cathepsin B, a lysosomal cysteine protease which functions as both a endopeptidase and a carboxypeptidase. Together with cathepsins D and L, cathepsin B is one of the most abundant proteases in the lysosome, and participates in intracellular protein degradation.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only