Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cathepsin B Sheep anti-Human, Polyclonal, Invitrogen™

Sheep Polyclonal Antibody

Manufacturer:  Invitrogen PA131403

Catalog No. PA131403


Add to cart

Description

Description

PA1-31403 detects Cathepsin B from human samples. PA1-31403 has been successfully used in Western blot applications. The PA1-31403 immunogen is: Human Cathepsin B purified from liver

Cathepsin B is a lysosomal cysteine protease that has been implicated in the pathogenesis ofrhematoid arthritis, muscular dystrophy and tumour metastasis.
Specifications

Specifications

Cathepsin B
Polyclonal
glycine buffered saline with 0.01% benzamidine, 0.1% EACA, 1mM EDTA and 0.09% sodium azide; pH 7.4
P07858
APPS, CPSB
Sheep
IgG
500 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1508
Immunodiffusion, Western Blot
Unconjugated
CTSB
Liquid
CTSB
Human Cathepsin B purified from liver
Ion-exchange chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit